Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May bind RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA-binding motif protein 41; RNA-binding protein 41
Gene Aliases: RBM41
UniProt ID: (Human) Q96IZ5
Entrez Gene ID: (Human) 55285
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support