Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MEKKKMVTQG NQEPTTTPDA MVQPFTTIPF PPPPQNGIPT EYGVPHTQDY AGQTGEHNLT LYGSTQAHGE QSSNSPSTQN GSLTTEGGAQ TDGQQSQTQS
This gene is one of several human genes similar to the C. elegans gene Fox-1. This gene encodes an RNA binding protein that is thought to be a key regulator of alternative exon splicing in the nervous system and other cell types. The protein binds to a conserved UGCAUG element found downstream of many alternatively spliced exons and promotes inclusion of the alternative exon in mature transcripts. The protein also interacts with the estrogen receptor 1 transcription factor and regulates estrogen receptor 1 transcriptional activity. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fox-1 homolog B; Fox-1 homolog Fxh; fox-1 homologue; Fyn-binding molecule 2; Hexaribonucleotide-binding protein 2; Repressor of tamoxifen transcriptional activity; RNA binding protein fox-1 homolog 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA-binding motif protein 9; RNA-binding protein 9
Gene Aliases: dJ106I20.3; Fbm2; Fox-2; FOX2; Fxh; HNRBP2; HRNBP2; RBFOX2; RBM9; RTA
UniProt ID: (Human) O43251, (Mouse) Q8BP71
Entrez Gene ID: (Human) 23543, (Mouse) 93686
Molecular Function:
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support