Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MGKVWKQQMY PQYATYYYPQ YLQAKQSLVP AHPMAPPSPS TTSSNNNSSS SSNSGWDQLS KTNLYIRGLP PHTTDQDLVK LCQPYGKIVS TKAILDKTTN KCKGYGFVDF DSPAAAQKAV SALKASGVQA QMAKQQEQDP TNLYISNLPL SMDEQELENM LKPFGQVIST RILRDSSGTS RGVGFARMES TEKCEAVIGH FNGKFIKTPP GVSAPTEPLL CKFADGGQKK RQNPNKYIPN GRPWHREGEV RLAGMTLTYD PTTAAIQNGF YPSPYSIATN RMITQTSITP YIASPVSAYQ VQSPSWMQPQ PYILQHPGAV LTPSMEHTMS LQPASMISPL AQQMSHLSLG STGTYMPATS AMQGAYLPQY AHMQTTAVPV EEASGQQQVA VETSNDHSPY TFQPNK
This gene encodes a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. Several transcript variants, resulting from alternative splicing and encoding different isoforms, have been described. A pseudogene for this locus is found on chromosome 12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-myc gene single strand binding protein 2; cervical cancer oncogene 4; RNA binding motif, single stranded interacting protein 1; RNA-binding motif, single-stranded-interacting protein 1; Single-stranded DNA-binding protein MSSP-1; suppressor of cdc 2 (cdc13) with RNA binding motif 2; Suppressor of CDC2 with RNA-binding motif 2
Gene Aliases: C2orf12; HCC-4; MSSP; MSSP-1; MSSP-2; MSSP-3; MSSP1; RBMS1; SCR2; YC1
UniProt ID: (Human) P29558
Entrez Gene ID: (Human) 5937
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support