Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPADLSGTW TLLSSDNFEG YMLALGIDFA TRKIAKLLKP QKVIEQNGDS FTIHTNSSLR NYFVKFKVGE EFDEDNRGLD NRKCKSLVIW DNDRLTCIQK GEKKNRGWTH WIEGDKLHLE MFCEGQVCKQ TFQRA (1-134 aa encoded by BC063013)
Due to its chemical instability and low solubility in aqueous solution, vitamin A requires cellular retinol-binding proteins (CRBPs), such as RBP7, for stability, internalization, intercellular transfer, homeostasis, and metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cellular retinoic acid-binding protein 4; Cellular retinoic acid-binding protein IV; cellular retinol binding protein 7; cellular retinol binding protein type III; CRABP-IV; CRABP4; CRBP4; cytosolic binding protein; putative cellular retinol-binding protein CRBP IV; retinoid binding protein 7; Retinoid-binding protein 7; retinol binding protein 7, cellular
Gene Aliases: 1110002J23Rik; CRBP-III; CRBP4; CRBPIV; RBP7; RGD1562168
UniProt ID: (Human) Q96R05, (Mouse) Q9EPC5
Entrez Gene ID: (Human) 116362, (Rat) 362662, (Mouse) 63954
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support