Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CBF-1; H-2K binding factor-2; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; RBP-J; RBP-J kappa; RBP-JK; rbp-jkappa; Recombining binding protein suppressor of hairless; Renal carcinoma antigen NY-REN-30; suppressor of hairless homolog
Gene Aliases: AI843960; AOS3; CBF1; csl; IGKJRB; IGKJRB1; Igkrsbp; KBF2; RBP-J; RBP-J kappa; RBPJ; RBPJK; RBPSUH; SUH
UniProt ID: (Human) Q06330, (Mouse) P31266
Entrez Gene ID: (Human) 3516, (Mouse) 19664
Molecular Function: DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support