Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNNGGKAEK ENTPSEANLQ EEEVRTLFVS GLPLDIKPRE LYLLFRPFKG YEGSLIKLTS KQPVGFVSFD SRSEAEAAKN ALNGIRFDPE IPQTLRLEFA KANTKMAKNK LVGTPNPSTP LPNTVPQFIA REPYELTVPA LYPSSPEVWA PYPLYPAELA PALPPPAFTY PASLHAQCFS PEAKPNTPVF CPLLQQIRFV SGNVFVTYQP TADQQRELPC (1-219 aa encoded by BC003608)
This gene encodes a member of the RRM family of RNA-binding proteins. The RRM domain is between 80-100 amino acids in length and family members contain one to four copies of the domain. The RRM domain consists of two short stretches of conserved sequence called RNP1 and RNP2, as well as a few highly conserved hydrophobic residues. The protein encoded by this gene has a single, putative RRM domain in its N-terminus. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heart and RRM expressed sequence; HEart, RRM Expressed Sequence; Hermes; RBP-MS; RNA binding protein gene with multiple splicing; RNA-binding protein gene with multiple splicing; RNA-binding protein with multiple splicing; RNA-binding protein with multiple splicing (RBP-MS)
Gene Aliases: 2010300K22Rik; 2700019M19Rik; AU017537; HERMES; RBP-MS; RBPMS; RGD1561067
UniProt ID: (Human) Q93062, (Mouse) Q9WVB0, (Rat) A0A8I6G705
Entrez Gene ID: (Human) 11030, (Mouse) 19663, (Rat) 498642
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support