Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAITQFRLFK FCTCLATVFS FLKRLICRSG RGRKLSGDQI TLPTTVDYSS VPKQTDVEEW TSWDEDAPTS VKIEGGNGNV ATQQNSLEQL EPDYFKDMTP TIRKTQKIVI KKREPLNFGI PDGSTGFSSR LAATQDLPFI HQSSELGDLD TWQENTNAWE EEEDAAWQAE EVLRQQKLAD REKRAAEQQR KKMEKEAQRL MKKEQNKIGV KLS
This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5'-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5' UTR, but encoding the same protein, have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAG9; cancer associated surface antigen; cancer associated surface antigen RCAS1; Cancer-associated surface antigen RCAS1; estrogen receptor binding fragment associated gene 9; Estrogen receptor-binding fragment-associated gene 9 protein; HGNC:3123; receptor binding cancer antigen expressed on SiSo cells; Receptor-binding cancer antigen expressed on SiSo cells
Gene Aliases: EB9; EBAG9; PDAF; RCAS1
UniProt ID: (Human) O00559
Entrez Gene ID: (Human) 9166
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support