Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
In the process of mRNA degradation, seems to play a role in mRNA decapping. Component of a complex containing DCP2 and DCP1A which functions in decapping of ARE-containing mRNAs. Promotes complex formation between DCP1A and DCP2. Enhances the catalytic activity of DCP2 (in vitro).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Autoantigen Ge-1; Autoantigen RCD-8; Enhancer of mRNA-decapping protein 4; Hedls; Human enhancer of decapping large subunit
Gene Aliases: BC022641; EDC4; Ge-1; GE1; HEDL5; HEDLS; RCD-8; RCD8
UniProt ID: (Human) Q6P2E9, (Mouse) Q3UJB9
Entrez Gene ID: (Human) 23644, (Mouse) 234699
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support