Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: QSNEETAGMV RHIYRDLEAA DAAALQAGNG EEEILPHCNL QVFTYTCDVG
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
RDH10 generates all-trans retinal from all-trans retinol and may plan an important role in the photic visual cycle. All-trans retinal is isomerized to 11-cis retinal by the retinal G protein-coupled receptor (RGR; MIM 600342) when the retinal pigment epithelium (RPE) is illuminated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Retinol dehydrogenase 10; Short chain dehydrogenase/reductase family 16C member 4; short chain dehydrogenase/reductase family 16C, member 4
Gene Aliases: RDH10; SDR16C4; UNQ9375/PRO34191
UniProt ID: (Human) Q8IZV5
Entrez Gene ID: (Human) 157506
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support