Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MEKCEAAAKD IRGETLNHHV NARHLDLASL KSIREFAAKI IEEEERVDIL INNAGVMRCP HWTTEDGFEM QFGVNHLGHF LLTNLLLDKL KASAPSRIIN LSSLAHVAGH IDFDDLNWQT RKYNTKAAYC QSKLAIVLFT KELSRRLQGS GVTVNALHPG VARTELGRHT GIHGSTFSST TLGPIFWLLV KSPELAAQPS TYLAVAEELA DVSGKYFDGL KQKAPAPEAE DEEVARRLWA ESARLVGLEA PSVREQPLPR
Does not exhibit retinol dehydrogenase (RDH) activity in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Retinol dehydrogenase 13; retinol dehydrogenase 13 (all-trans and 9-cis); Short chain dehydrogenase/reductase family 7C member 3; short chain dehydrogenase/reductase family 7C, member 3
Gene Aliases: PSEC0082; RDH13; SDR7C3; UNQ736/PRO1430
UniProt ID: (Human) Q8NBN7
Entrez Gene ID: (Human) 112724
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support