Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MIFALLPKPR KNVAGEIVLI TGAGSGLGRL LALQFARLGS VLVLWDINKE GNEETCKMAR EAGATRVHAY TCDCSQKEGV YRVADQVKKE VGDVSILINN AGIVTGKKFL DCPDELMEKS FDVNFKAHLW TYKAFLPAMI ANDHGHLVCI SSSAGLSGVN GLADYCASKF AAFGFAESVF VETFVQKQKG IKTTIVCPFF IKTGMFEGCT TGCPSLLPIL EPKYAVEKIV EAILQEKMYL YMPKLLYFMM FLKSFLPLKT GLLIADYLGI LHAMDGFVDQ KKKL
RDHE2 belongs to a family of short-chain alcohol dehydrogenases/reductases that catalyze the first and rate-limiting step that generates retinaldehyde from retinol (Matsuzaka et al., 2002 [PubMed 12372410]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EPHD-2; epidermal retinal dehydrogenase 2; Epidermal retinol dehydrogenase 2; RDH-E2; retinal short chain dehydrogenase reductase; retinal short chain dehydrogenase reductase (Rdhe2); retinal short chain dehydrogenase reductase 2; Retinal short-chain dehydrogenase reductase 2; retSDR2; short chain dehydrogenase reductase 9; Short-chain dehydrogenase reductase 9; Short-chain dehydrogenase/reductase family 16C member 5
Gene Aliases: EPHD-2; RDH#2; RDH-E2; RDHE2; retSDR2; RGD1565999; Scdr9; SDR16C5
UniProt ID: (Human) Q8N3Y7, (Mouse) Q7TQA3
Entrez Gene ID: (Human) 195814, (Mouse) 242285, (Rat) 297805
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support