Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total REC8 protein.
This gene encodes a member of the kleisin family of SMC (structural maintenance of chromosome) protein partners. The protein localizes to the axial elements of chromosomes during meiosis in both oocytes and spermatocytes. In the mouse, the homologous protein is a key component of the meiotic cohesion complex, which regulates sister chromatid cohesion and recombination between homologous chromosomes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cohesin Rec8p; cohesion rec8p; human homolog of rad21, S. pombe; kleisin-alpha; meiotic recombination and sister chromatid cohesion phosphoprotein of the rad21p family; Meiotic recombination protein REC8 homolog; meiotic recombination protein REC8-like 1; REC8 homolog; REC8-like 1; recombination and sister chromatid cohesion protein homolog
Gene Aliases: HR21spB; REC8; REC8L1; Rec8p
UniProt ID: (Human) O95072
Entrez Gene ID: (Human) 9985
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support