Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
DDIT4 gene ontology annotations related to this gene include cellular response to dexamethasone stimulus; intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator; negative regulation of glycolytic process; negative regulation of TOR signaling; protein-containing complex disassembly; reactive oxygen species metabolic process; response to hypoxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA damage-inducible transcript 4 protein; DNA-damage-inducible transcript 4; HIF-1 responsive protein RTP801; Protein regulated in development and DNA damage response 1; REDD-1; RP11-442H21.1
Gene Aliases: DDIT4; Dig2; REDD-1; REDD1; RTP801
UniProt ID: (Human) Q9NX09
Entrez Gene ID: (Human) 54541
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support