Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
REEP1 belongs to the DP1 family and may enhance the cell surface expression of odorant receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: rcG_56072; Receptor expression-enhancing protein 1; Spastic paraplegia 31 protein
Gene Aliases: C2orf23; D6Ertd253e; HMN5B; REEP1; RGD1305230; SPG31; Yip2a
UniProt ID: (Human) Q9H902, (Mouse) Q8BGH4
Entrez Gene ID: (Human) 65055, (Mouse) 52250, (Rat) 362384
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support