Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) RELM beta or rmResistin.
Reconstitute at 0.5 mg/mL in sterile PBS.
RELM alpha, also known as Resistin-like alpha or FIZZ1, is a small cytokine (88 aminoacids, MW 9.4 kDa) produced mostly by macrophages. Its highest constitutive expression has been observed in small peritoneal macrophages, alveolar macrophages and adipose tissue macrophages. It is strongly induced by IL-4 and IL-13. Together with Arginase 1 and Ym-1, RELM alpha is considered one of the hallmarks of the M2 polarization of mouse macrophages, however, macrophages polarized with IL-10 (M2c) express neither RELM alpha nor Arginase 1. Also, the expression patterns of RELM alpha and Arginase 1 in untreated tissue macrophages significantly differ. Elevated levels of RELM alpha may indicate parasitic infection. Humans do not have RELM alpha gene but human Resistin resembles mouse RELM alpha in respect to sequence, tissue distribution and function. Other members of RELM family in mouse: RELM beta, RELM gamma and Resistin display different tissue distribution and functional properties. RELM alpha plays an important role in parasite infection, mucosal inflammation and allergy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cysteine-rich secreted protein A12-gamma; Cysteine-rich secreted protein FIZZ1; FIZZ2; found in inflammatory zone 1; HXCP2; Hypoxia-induced mitogenic factor; Parasite-induced macrophage novel gene 1 protein; RELM-beta; RELMalpha; RELMb; Resistin-like alpha
Gene Aliases: 1810019L16Rik; Fizz-1; Fizz1; HIMF; Pmng1; RELM-alpha; RELMa; RELMalpha; Retnla; Xcp2
UniProt ID: (Mouse) Q9EP95
Entrez Gene ID: (Mouse) 57262
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support