Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LMVPRLEHVV EEEKVDISED GMKALVTLSS GDMRRALNIL QSTNMAFGKV TEETVYTCTG HPLKSDIANI LDWMLNQDFT TAYRNITELK TLKGLALHDI LTEIHLFVHR VDFPSSVRIH LLTKMADIEY RLSVGTNEKI QLSSLIAAFQ VTRDLIVAEA; Positive Samples: HeLa, U-251MG, SKOV3, MCF-7, Mouse liver, Mouse spleen; Cellular Location: Nucleus
The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kD. This gene encodes the 36 kD subunit. This subunit can interact with the C-terminal region of PCNA. It forms a core complex with the 38 and 40 kDa subunits. The core complex possesses DNA-dependent ATPase activity, which was found to be stimulated by PCNA in an in vitro system. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A1 36 kDa subunit; Activator 1 36 kDa subunit; Activator 1 subunit 5; MGC1155; replication factor C (activator 1) 5, 36.5kDa; Replication factor C 36 kDa subunit; replication factor C 5; Replication factor C subunit 5; replication factor C, 36 kDa; replication factor C, 36-kDa subunit; RF-C 36 kDa subunit; RFC, 36.5 kD subunit; RFC36
Gene Aliases: 2610020K06Rik; 2610209F07Rik; 36.5kDa; 36kDa; Recc5; RFC36; RFC5
UniProt ID: (Human) P40937, (Mouse) Q9D0F6
Entrez Gene ID: (Human) 5985, (Rat) 304528, (Mouse) 72151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support