Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TEAEIICDVV CLVYDVSNPK SFEYCARIFK QHFMDSRIPC LIVAAKSDLH EVKQEYSISP TDFCRKHKMP PPQAFTCNTA DAPSKDIFVK LTTMAMYP
Mitochondrial GTPase involved in mitochondrial trafficking. RHOT1 is probably involved in control of anterograde transport of mitochondria and their subcellular distribution.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MIRO-1; mitochondrial Rho (MIRO) GTPase 1; Mitochondrial Rho GTPase 1; rac-GTP binding protein-like protein; Rac-GTP-binding protein-like protein; Ras homolog gene family member T1; ras homolog gene family, member T1
Gene Aliases: ARHT1; MIRO-1; MIRO1; RHOT1
UniProt ID: (Human) Q8IXI2
Entrez Gene ID: (Human) 55288
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support