Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
RIAM appears to function in the signal transduction from Ras activation to actin cytoskeletal remodeling. Suppresses insulin-induced promoter activities through AP1 and SRE. Mediates Rap1-induced adhesion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amyloid beta (A4) precursor protein-binding, family B, member 1 interacting protein; Amyloid beta A4 precursor protein-binding family B member 1-interacting protein; APBB1-interacting protein 1; PREL-1; proline rich EVH1 ligand 1; Proline-rich EVH1 ligand 1; Proline-rich protein 48; Proline-rich protein 73; Rap1-GTP-interacting adapter molecule; Rap1-GTP-interacting adaptor molecule; Rap1-interacting adaptor molecule; RARP-1; Retinoic acid-responsive proline-rich protein 1; RIAM; RIAMRARP-1
Gene Aliases: 9930118P07Rik; APBB1IP; INAG1; PREL1; Prp48; RARP1; RIAM
UniProt ID: (Human) Q7Z5R6, (Mouse) Q8R5A3
Entrez Gene ID: (Human) 54518, (Mouse) 54519
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support