Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AVO3 homolog; DKFZp686B11164; hAVO3; mAVO3; MGC39830; pianissimo; Protein pianissimo; Rapamycin-insensitive companion of mTOR; RICTOR; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3
Gene Aliases: 4921505C17Rik; 6030405M08Rik; AVO3; AW492497; D530039E11Rik; hAVO3; KIAA1999; PIA; RICTOR
UniProt ID: (Human) Q6R327, (Mouse) Q6QI06
Entrez Gene ID: (Human) 253260, (Mouse) 78757
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support