Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen Sequence: FKPRNYQLEL ALPAMKGKNT IICAPTGCGK TFVSLLICEH HLKKFPQGQK GKVVFFANQI PVYEQQKSVF SKYFERHGYR VTGISGATAE NVPVEQIVEN NDIIILTPQI LVNNLKKGTI PSLSIFTLMI FDECHNTSKQ HPYNMIMFNY LDQKLGGSSG PLPQVIGLTA SVGVGDAKNT DEALDYICKL CASLDASVIA TVKHNLEELE QVVYK
Retinoic acid-inducible gene I, RIG-I is a pattern recognition receptor (PRR) involved in the recognition of viral dsRNA. Along with MDA5, RIG-I detects viral dsRNA and activates the innate immune response. Both MDA5 and RIG-I are RNA helicases and they perform overlapping as well as distinct roles. RIG-I is activated by dsRNAs without a 5'-triphosphate end and short dsRNAs, whereas MDA5 is activated by long dsRNAs. Once activated, both proteins signal through IPS-1 activating transcription factors NF-kappaB and IRF-3 (1) and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus (3), paromyxoviruses, Japanese encephalitis virus, and West Nile virus. MicroRNA-146a has been implicated in feedback inhibition of RIG-I-dependant antiviral response by negatively regulating RIG-I targets TRAF6, IRAK1, and IRAK2. Recent evidence has implicated RIG-I in the detection of cytosolic DNA through RNA polymerase III activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antiviral innate immune response receptor RIG-I; ATP-dependent RNA helicase DDX58; DEAD (Asp-Glu-Ala-Asp) box polypeptide 58; DEAD box protein 58; DEAD-box protein 58; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; DEAD/H box polypeptide RIG-I; probable ATP-dependent RNA helicase DDX58; Retinoic acid-inducible gene 1 protein; Retinoic acid-inducible gene I protein; retinoic acid-inducible gene-I; RIG-1; RIG-I; RIG-I-like receptor 1; RLR-1; RNA helicase RIG-I; RNA sensor RIG-I
Gene Aliases: 6430573D20Rik; C330021E21; DDX58; RIG-I; RIGI; RLR-1; SGMRT2
UniProt ID: (Human) O95786, (Mouse) Q6Q899
Entrez Gene ID: (Human) 23586, (Mouse) 230073
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support