Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Nuclear receptor interacting protein 1 is a nuclear protein that specifically interacts with the hormone-dependent activation domain AF2 of nuclear receptors. Also known as RIP140, this protein modulates transcriptional activity of the estrogen receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERAP 140; FLJ77253; nrip1; Nuclear factor RIP140; Nuclear receptor-interacting protein 1; OTTHUMP00000083036; OTTHUMP00000083037; Receptor-interacting protein 140
Gene Aliases: 6030458L20Rik; 8430438I05Rik; 9630050P12; AA959574; AW456757; NRIP1; RIP140
UniProt ID: (Human) P48552, (Mouse) Q8CBD1
Entrez Gene ID: (Human) 8204, (Rat) 304157, (Mouse) 268903
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support