Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Binds and exchanges GTP and GDP. Binds and modulates the activation of POU4F1 as gene expression regulator.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein Rit2; GTP-binding protein Roc2; Ras-like protein expressed in neurons; Ras-like without CAAX 2; Ras-like without CAAX protein 2; Ric-like, expressed in neurons
Gene Aliases: RIBA; RIN; RIT2; ROC2
UniProt ID: (Human) Q99578
Entrez Gene ID: (Human) 6014
Molecular Function:
small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support