Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MSEGVGTFRM VPEEEQELRA QLEQLTTKDH GPVFGPCSQL PRHTLQKAKD ELNEREETRE EAVRELQEMV QAQAASGEEL AVAVAERVQE KDSGFFLRFI RARKFNVGRA YELLRGYVNF RLQYPELFDS LSPEAVRCTI EAGYPGVLSS RDKYGRVVML FNIENWQSQE ITFDEILQAY CFILEKLLEN EETQINGFCI IENFKGFTMQ QAASLRTSDL RKMVDMLQDS FPARFKAIHF IHQPWYFTTT YNVVKPFLKS KLLERVFVHG DDLSGFYQEI DENILPSDFG GTLPKYDGKA VAEQLFGPQA QAENTAF
Cellular retinaldehyde-binding protein (CRALBP) plays an important role in the regeneration of 11-cis-retinal for use in rod visual pigments such as opsin and rhodopsin. Once 11-cis-retinal is photoisomerized in the rod outer segment, it is converted to all-trans-retinal and further modified into all-trans-retinol. All-trans-retinol then diffuses into the retinal pigment epithelium (RPE) to be converted back to 11-cis-retinol and further oxidized into 11-cis-retinal (both by CRALBP). Genetic mutations involving CRALBP's lack of function have been linked to visual disease such as bothnia dystrophy, retinitis punctata albescens, retina pigmentosa, and Newfoundland rod-cone dystrophy. The presence of CRALBP serves as a marker for RPE and Muller glial cells of the retina.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cellular retinaldehyde-binding protein; cellular retinaldehyde-binding protein-1; Retinaldehyde-binding protein 1
Gene Aliases: CRALBP; RLBP1
UniProt ID: (Human) P12271
Entrez Gene ID: (Human) 6017
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support