Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSDAAGDIV DIREEIKCDF EFKAKHRIAH KPHSKPKTSD IFEADIANDV KSKDLLADKE LWARLEELER QEELLGELDS KPDTVIANGE DTTSSEEEKE DRNTNVNAMH QVTDSHTPCH KDVASSEPFS GQVNSQLNCS VNGSSSYHSD DDDDDDDDDD DDNIDDDDGD NDHEALGVGD NSIPTIYFSH TVEPKRVRIN TGKNTTLKFS EKKEEAKRKR KNSTGSGHSA QELPTIRTPA DIYRAFVDVV NGEYVPRKSI LKSRSRENSV CSDTSESSAA EFDDRRGVLR SISCEEATCS DTSESILEEE PQENQKKLLP LSVT (125-448 aa encoded by B C026184)
Involved in gene transcription regulation. Acts as a transcriptional repressor in concert with the corepressor UXT to regulate androgen receptor (AR) transcription. May act as a tumor suppressor to repress AR-mediated gene transcription and to inhibit anchorage-independent growth in prostate cancer cells. Required for cell survival in ovarian cancer cells. Together with UXT, associates with chromatin to the NKX3-1 promoter region. Antagonizes transcriptional modulation via hepatitis B virus X protein. Plays a central role in maintaining S6K1 signaling and BAD phosphorylation under normal growth conditions thereby protecting cells from potential deleterious effects of sustained S6K1 signaling. The URI1-PPP1CC complex acts as a central component of a negative feedback mechanism that counteracts excessive S6K1 survival signaling to BAD in response to growth factors. Mediates inhibition of PPP1CC phosphatase activity in mitochondria. Coordinates the regulation of nutrient-sensitive gene expression availability in a mTOR-dependent manner. Seems to be a scaffolding protein able to assemble a prefoldin-like complex that contains PFDs and proteins with roles in transcription and ubiquitination. [Uniprot]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10575; Protein NNX3; Protein phosphatase 1 regulatory subunit 19; protein phosphatase 1, regulatory subunit 19; RNA polymerase II subunit 5-mediating protein; RPB5-mediating protein; Unconventional prefoldin RPB5 interactor; Unconventional prefoldin RPB5 interactor 1; URI1 prefoldin-like chaperone
Gene Aliases: C19orf2; C80913; NNX3; PPP1R19; RMP; URI; URI1
UniProt ID: (Human) O94763, (Mouse) Q3TLD5
Entrez Gene ID: (Human) 8725, (Mouse) 19777
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support