Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detected 30 kDa band with Testis and L02 cell lysate.
Immunogen sequence: METFPLLLL SLGLVLAEAS ESTMKIIKEE FTDEEMQYDM AKSGQEKQTI EILMNPILLV KNTSLSMSKD DMSSTLLTFR SLHYNDPKGN SSGNDKECCN DMTVWRKVSE ANGSCKWSNN FIRSSTEVMR RVHRAPSCKF VQNPGISCCE SLELENTVCQ FTTGKQFPRC QYHSVTSLEK ILTVLTGHSL MSWLVCGSKL (1-199 aa encoded by BC025410)
RNASE11 (Ribonuclease A Family Member 11 (Inactive)) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and ribonuclease activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein Li 84p; Probable ribonuclease 11; ribonuclease 11; ribonuclease A J1; ribonuclease, RNase A family, 11 (non-active); RNase 11
Gene Aliases: C14orf6; HEL-S-84p; RAJ1; RNASE11; UNQ5832/PRO19669
UniProt ID: (Human) Q8TAA1
Entrez Gene ID: (Human) 122651
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support