Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDLSELERD NTGRCRLSSP VPAVCRKEPC VLGVDEAGRG PVLGPMVYAI CYCPLPRLAD LEALKVADSK TLLESERERL FAKMEDTDFV GWALDVLSPN LISTSMLGRV KYNLNSLSHD TATGLIQYAL DQGVNVTQVF VDTVGMPETY QARLQQSFPG IEVTVKAKAD ALYPVVSAAS ICAKVARDQA VKKWQFVEKL QDLDTDYGSG YPNDPKTKAW LKEHVEPVFG FPQFVRFSWR TAQTILEKEA EDVIWEDSAS ENQEGLRKIT SYFLNEGSQA RPRSSHRYFL ERGLESATSL (1-299 aa encoded by BC011748)
Ribonucleases (RNAses) H are enzymes that hydrolyze the RNA strands of RNA/DNA hybrids. The major role of these enzymes is to remove the RNA strand from the RNA/DNA hybrids that form during DNA replication and repair. RNAse H2 is made up of three subunits; all three are required for RNAse activity. Recent evidence has demonstrated that mutations in RNAse H2A or any of the other subunits result in Aicardi-Goutieres syndrome (AGS), a neurological disorder with similar symptoms to viral brain infections including high levels of IFN-alpha in the cerebral spinal fluid. Similar conditions are observed with mutations in TREX1, a single-stranded DNA exonuclease, suggesting that RNAse H2 and TREX1 may have similar roles, and that mutations in any of these genes lead to an accumulation of intracellular nucleic acids, triggering an inflammatory response through activation of the innate immune system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGS4; Aicardi-Goutieres syndrome 4 protein; Ribonuclease H2 subunit A; ribonuclease H2, large subunit; ribonuclease H2, subunit A; ribonuclease HI; Ribonuclease HI large subunit; Ribonuclease HI subunit A; ribonuclease HI, large subunit; RNase H(35); RNase H2 subunit A; RNase HI large subunit
Gene Aliases: 2400006P09Rik; AGS4; JUNB; RNASEH2A; RNASEHI; RNHIA; RNHL
UniProt ID: (Human) O75792, (Mouse) Q9CWY8
Entrez Gene ID: (Human) 10535, (Mouse) 69724
Molecular Function:
endoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support