Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KTFCEGSPR AQREVSCLQS SRENSHLVTF YGSESHRGHL FVCVTLCEQT LEACLDVHRG EDVENEEDEF AQNVLSSIFK AVQELHLSCG YTHQDLQPQN ILIDSKKAAH LADFDKSIKW AGDPQEVKRD LEDLGRLVLY VVKKGSISFE ELKAQSNEEV VQLSPDEETK DLIHRLFHPG EHVRDCLSDL LGHPFFWTWE SRYRTLRNVG NESDIKTRKS ESEILRLLQP GPSEHSKSFD KWTTKINECV MKKMNKFYEK RGNFYQNTVG DLLKFIRNLG EHIDEEKHKK MKLKIGDPSL YFQKTFPDLV IYVYTKLQNT EYRKHFPQTH SPNKPQCDGA GGASGLASPG C (392-741 aa encoded by BC115697)
A virally induced endoribonuclease, RNase L is the terminal factor of the anti-viral action of interferon. Activation of RNase L results in inhibition of viral proliferation. Activation of RNase L has also been determined to degrade RNA, thus, initiating a cellular stress response, inducing apoptosis. Since mutations in RNase L relate to an increased incidence of prostate cancer, the activation of RNase L functions in counteracting prostate cancer, via apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2',5'-oligoisoadenylate synthetase-dependent; 2-5A-dependent ribonuclease; 2-5A-dependent RNAase; 2-5A-dependent RNase; interferon-induced 2-5A-dependent RNase; Ribonuclease 4; Ribonuclease L; ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent); RNase L
Gene Aliases: E230029I04Rik; PRCA1; RNASEL; RNS4
UniProt ID: (Human) Q05823, (Mouse) Q05921
Entrez Gene ID: (Human) 6041, (Mouse) 24014
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support