Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MAEQLSPGKA VDQVCTFLFK KPGRKGAAGR RKRPACDPEP GESGSSSDEG CTVVRPEKKR VTHNPMIQKT RDSGKQKAAY GDLSSEEEEE NEPESLGVVY KSTRSAKPVG PEDMGATAVY ELDTEKERDA QAIFERSQKI QEELRGKEDD KIYRGINNYQ KYMKPKDTSM GNASSGMVRK GPIRAPEHLR ATVRWDYQPD ICKDYKETGF CGFGDSCKFL HDRSDYKHGW QIERELDEGR YGVYEDENYE VGSDDEEIPF KCFICRQSFQ NPVVTKCRHY FCESCALQHF RTTPRCYVCD QQTNGVFNPA KELIAKLEKH RATGEGGASD LPEDPDEDAI PIT
This intronless gene encodes a protein which contains a C3H1-type zinc finger domain and a C3HC4 Ring-type (Really Interesting New Gene-type) zinc finger domain. The Ring-type zinc finger domain is identified in various tumor suppressors, DNA repair genes and cytokine receptor-associated molecules, and is probably involved in mediating protein-protein interactions. [provided by RefSeq, May 2010].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cwc24 homolog; E3 ubiquitin-protein ligase RNF113A; RING finger protein 113A; Zinc finger protein 183; zinc finger protein 183 (RING finger, C3HC4 type)
Gene Aliases: Cwc24; RNF113; RNF113A; TTD5; ZNF183
UniProt ID: (Human) O15541
Entrez Gene ID: (Human) 7737
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support