Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RNF113B (Ring Finger Protein 113B) is a Protein Coding gene. An important paralog of this gene is RNF113A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RING finger protein 113B; Zinc finger protein 183-like 1
Gene Aliases: bA10G5.1; RNF113B; RNF161; ZNF183L1
UniProt ID: (Human) Q8IZP6
Entrez Gene ID: (Human) 140432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support