Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGSVLSTDS GKSAPASATA RALERRRDPE LPVTSFDCAV CLEVLHQPVR TRCGHVFCRS CIATSLKNNK WTCPYCRAYL PSEGVPATDV AKRMKSEYKN CAECDIVLYL SEMRAHIRTC QKYIDKYGPL QELEETAARC VCPFCQRELY EDSLLDHCIT HHRSERRPVF CPLCRLIPDE NPSSFSGSLI RHLQVSHTLF YDDFIDFNII EEALIRRVLD RSLLEYVNHS NTA (1-232 aa encoded by BC012021)
TRAC-1 is a 232 amino acid protein containing an N-terminal RING-type zinc finger domain. It functions as a positive regulator in the T-cell receptor signaling pathway, thus leading to T-cell activation. It mediates ubiquitination and subsequent proteasomal degradation of target proteins. TRAC-1 is expressed in lymphoid tissues, including bone marrow, spleen, thymus as well as in other tissues. It is predominantly expressed in the CD4+ and CD8+ T-cells, suggesting that it is preferentially confined to T-cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF125; RING finger protein 125; ring finger protein 125, E3 ubiquitin protein ligase; T-cell RING activation protein 1; T-cell ring protein identified in activation screen; TRAC-1
Gene Aliases: 4930553F04Rik; RNF125; TNORS; TRAC-1; TRAC1
UniProt ID: (Human) Q96EQ8, (Mouse) Q9D9R0
Entrez Gene ID: (Human) 54941, (Rat) 361296, (Mouse) 67664
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support