Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NTGDIQKYL TPKSQFGSAS HSEAVQEVRK DSVSKDIDSS DRKSPTGQDT EIEDMPTLSP QISLGVGEQG ADSSIESPMP WLCACGAEWY HEGNVKTRPS NHGKELCVLS HERPKTRVPY SKETAVMPCG RTESGCAPTS GVTQTNGNNT GETENEESCL LISKEISKRK NQESSFEAVK DPCFSAKRRK VSPESSPDQE ETEINFTQKL IDLEHLLFER HKQEEQDRLL ALQLQKEVDK EQMVPNRQKG SPDEYHLRAT SSPPDKVLNG QRKNPKDGNF KRQTHTKHPT PERGSRDKNR QVSLKMQLKQ SVNRRKMPNS TRDHCKVSKS AHSLQPSISQ KSVFQMFQRC TK (221-571 aa encoded by BC033791)
RNF168 was identified as a chromatin-associated RING finger protein that acts as a ubiquitin ligase both in vitro and in vivo. RNF168 targets histones H2A and H2AX, but not H2B, forming K63 polyubiquitin chains. Upon formation of DNA double strand breaks, RNF168 is recruited to the site of DNA damage where it co-localizes with -gammaH2AX and 53BP1 in an RNF8-dependent manner. This localization of RNF168 increases the local concentration of ubiquinated proteins to the threshold required for retention of the proteins 53BP1 and BRCA1, facilitating the downstream signaling cascade. Thus, RNF168 defines a new pathway demonstrating a functional cooperation between E3 ligases in genome maintenance. At least three isoforms of RNF168 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF168; FLJ35794; hRNF168; RING finger protein 168; ring finger protein 168, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF168
Gene Aliases: hRNF168; RNF168
UniProt ID: (Human) Q8IYW5
Entrez Gene ID: (Human) 165918
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support