Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSQAVQTNG TQPLSKTWEL SLYELQRTPQ EAITDGLEIV VSPRSLHSEL MCPICLDMLK NTMTTKECLH RFCADCIITA LRSGNKECPT CRKKLVSKRS LRPDPNFDAL ISKIYPSRDE YEAHQERVLA RINKHNNQQA LSHSIEEGLK IQAMNRLQRG KKQQIENGSG AEDNGDSSHC SNASTHSNQE AGPSNKRTKT SDDSGLELDN NNAAMAIDPV MDGASEIELV FRPHPTLMEK DDSAQTRYIK TSGNATVDHL SKYLAVRLAL EELRSKGESN QMNLDTASEK QYTIYIATAS GQFTVLNGSF SLELVSEKYW KVNKPMELYY APTKEHK (1-336 aa encoded by BC012583)
RNF2 (E3 ubiquitin-protein ligase RING2) is a ligase that mediates monoubiquitination of Lys-119 of histone H2A (H2AK119Ub). Therefore, it plays a central role in histone code and gene regulation. The ligase activity is enhanced by BMI1/PCGF4. H2AK119Ub gives a specific tag for epigeneti transcriptional repression and participates in X chromosome inactivation of female mammals. RNF2 may be involved in the initiation of both imprinted and random X inactivation. RNF2 is also an essential component of a Polycomb group (PcG) multiprotein PRC1-like complex. This complex class is required to maintain the transcriptionally repressive state of many genes including Hox genes throughout development. The PcG PRC1 complex acts via chromatin remodeling and modification of histones and rendering chromatin heritably changed in its expressibility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAP 1; DinG protein; E3 ubiquitin-protein ligase RING2; GS1-120K12.1; HIP2-interacting protein 3; HIPI 3; Huntingtin-interacting protein 2-interacting protein 3; OTTHUMP00000060668; Protein DinG; RING 1B; RING 2; RING finger protein 1B; RING finger protein 2; RING finger protein BAP-1; RING-type E3 ubiquitin transferase RING2; RING1b; RNF 2
Gene Aliases: AI326319; AI450156; AU019207; BAP-1; BAP1; DING; HIPI3; RING1B; RING2; RNF2
UniProt ID: (Human) Q99496, (Mouse) Q9CQJ4
Entrez Gene ID: (Human) 6045, (Rat) 304850, (Mouse) 19821
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support