Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: IKSSVSTKPH GCLLPPHSSA PDRLESMEVD LSPSPIRKSE IAAGPARISM ISPPQDGRMA PCARRVCHFQ RFTMFLHRRL SSLAAPPSVQ FWKARGTHQL
This gene encodes a RING finger protein that may function as a ubiquitin ligase. The encoded protein may be involved in meiotic recombination. This gene is located within a linkage disequilibrium block and polymorphisms in this gene may influence recombination rates. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Probable E3 SUMO-protein ligase RNF212; Probable E3 SUMO-protein transferase RNF212; RING finger protein 212
Gene Aliases: RNF212; ZHP3
UniProt ID: (Human) Q495C1
Entrez Gene ID: (Human) 285498
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support