Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: KPLWHMFKPP PALECRRCHI KCHKDHMDKK EEIIAPCKVY YDISTAKNLL LLANSTEEQQ KWVSRLVKKI PKKPPAPDPF ARSSPRTSMK IQQNQSIRRP SRQLAPNKPS
ROCK2, the predominant ROCK isoform in skeletal muscle, is an important regulator downstream of Rho GTPases. ROCK2 regulates cytokinesis, smooth muscle contraction, the formation of actin stress fibers and focal adhesions, and the activation of the c-fos serum response element. This protein, which is an isozyme of ROCK1 is a target for the small GTPase Rho.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p164 ROCK-2; Rho kinase 2; Rho-associated protein kinase 2; Rho-associated, coiled-coil-containing protein kinase 2; Rho-associated, coiled-coil-containing protein kinase II; ROCK II; ROCK-II; rockii
Gene Aliases: KIAA0619; ROCK-II; ROCK2
UniProt ID: (Human) O75116
Entrez Gene ID: (Human) 9475
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support