Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPLPDTMFC AQQIHIPPEL PDILKQFTKA AIRTQPADVL RWSAGYFSAL SRGDPLPVKD RMEMPTATQK TDTGLTQGLL KVLHKQCHHK RYVELTDLEQ KWKNLCLPKE KFKALLQLDP CENKIKWINF LALGCSMLGG SLNTALKHLC EILTDDREGG PARIPFKTFS YVYRYLARLD SDVSPLETES YLASLKENID ARKNGMIGLS DFFFPKRKLL ESIENSEDVG H (1-230 aa encoded by BC014607)
Principal Names: ROPN1L; Ropporin-1 like; ASP; AKAP-associated sperm protein; Official Gene Symbol: ROPN1L Gene ID: 83853 (human) Gene Map Locus: 5p15.2 (human) AKAPs act as scaffolding proteinscoordinating phosphatase/kinase signaling at the cytoskeleton. They interact with PKAs and anchor them to specific sub-cellular compartments and participate in multiple cellular processes including growth, metabolism and differentiation. ASP, a 26 kDa protein, is a novel AKAP-interacting protein. It interacts with the amphipathic helix of AKAP3 and facilitates AKAP3-PKA interactions. It consists of a conserved N-terminal AKAP-binding domain and a central cysteine-rich Fz domain. It is specifically expressed in the sperms and might play a significant role in sperm motility and also facilitate cytoskeletal reorganization during spermiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-kinase anchoring protein-associated sperm protein; AKAP-associated sperm protein; radial spoke head 11 homolog; ROPN1-like protein; ropporin 1-like; Ropporin-1-like protein
Gene Aliases: Akapasp; ASP; AV047578; ROPN1L; RSPH11
UniProt ID: (Human) Q96C74, (Mouse) Q9EQ00
Entrez Gene ID: (Human) 83853, (Rat) 685646, (Mouse) 252967
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support