Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRAADSGSWE RVRQLAAQGE PAPSCGAGAG PARPPGPAAC EQCVDAAGPG DRPRAGVPRV RADGDCSQPV LLREEVSRLQ EEVHLLRQMK EMLAKDLEES QGGKSSEVLS ATELRVQLAQ KEQELARAKE ALQAMKADRK RLKGEKTDLV SQMQQLYATL ESREEQLRDF IRNYEQHRKE SEDAVKALAK EKDLLEREKW ELRRQAKEAT DHATALRSQL DLKDNRMKEL EAELAMAKQS LATLTKDVPK RHSLAMPGET VLNGNQEWVV QADLPLTAAI RQSQQTLYHS HPPHPADRQA VRVSPCHSRQ PSVISDASAA EGDRSSTPSD INSPRHRTHS LCNGDSPGPV QKNLHNPIVQ SLEDLEDQKR KKKKEKMGFG SISRVFARGK QRKSLDPGLF DGTAPDYYIE EDADW
Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kazrin
Gene Aliases: C1orf196; HRIHFB2003; KAZ; KAZN; KIAA1026
UniProt ID: (Human) Q674X7, (Rat) Q5FWS6
Entrez Gene ID: (Human) 23254, (Rat) 313672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support