Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRPPACWWLL APPALLALLT CSLAFGLASE DTKKEVKQSQ DLEKSGISRK NDIDLKGIVF VIQSQSNSFH AKRAEQLKKS ILKQAADLTQ ELPSVLLLHQ LAKQEGAWTI LPLLPHFSVT YSRNSSWIFF CEEETRIQIP KLLETLRRYD PSKEWFLGKA LHDEEATIIH HYAFSENPTV FKYPDFAAGW ALSIPLVNKL TKRLKSESLK SDFTIDLKHE IALYIWDKGG GPPLTPVPEF CTNDVDFYCA TTFHSFLPLC RKPVKKKDIF VAVKTCKKFH GDRIPIVKQT WESQASLIEY YSDYTENSIP TVDLGIPNTD RGHCGKTFAI LERFLNRSQD KTAWLVIVDD DTLISISRLQ HLLSCYDSGK PVFLGERYGY GLGTGGYSYI TGGGGMVFSR EAVRRLLASK CRCYSNDAPD DMVLGMCFSG LGIPVTHSPL FHQARPVDYP KDYLSHQVPI SFHKHWNIDP VKVYFTWLAP SDEDKARQET QKGFREEL
The protein encoded by this gene is a beta-1, 3-glucosyltransferase that transfers glucose to O-linked fucosylglycans on thrombospondin type-1 repeats (TSRs) of several proteins. The encoded protein is a type II membrane protein. Defects in this gene are a cause of Peters-plus syndrome (PPS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta 1,3-galactosyltransferase-like; Beta 3-glucosyltransferase; beta 3-glycosyltransferase-like; Beta-1,3-glucosyltransferase; Beta-3-glycosyltransferase-like; Beta3Glc-T; UDP-GAL:beta-GlcNAc beta-1,3-galactosyltransferase-like
Gene Aliases: B3GALTL; B3Glc-T; B3GLCT; B3GTL; beta3Glc-T; Gal-T
UniProt ID: (Human) Q6Y288
Entrez Gene ID: (Human) 145173
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support