Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
The RP2 locus has been implicated as one cause of X-linked retinitis pigmentosa. The predicted gene product shows homology with human cofactor C, a protein involved in the ultimate step of beta-tubulin folding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIAA0215; Protein XRP2
Gene Aliases: AI662636; DELXp11.3; NM23-H10; NME10; RGD1565124; RP2; Rp2h; TBCCD2; XRP2
UniProt ID: (Human) O75695, (Mouse) Q9EPK2
Entrez Gene ID: (Human) 6102, (Rat) 367714, (Mouse) 19889
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support