Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQRPGPFST LYGRVLAPLP GRAGGAASGG GGNSWDLPGS HVRLPGRAQS GTRGGAGNTS TSCGDSNSIC PAPSTMSKAE EAKKLAGRAA VENHVRNNQV LGIGSGSTIV HAVQRIAERV KQENLNLVCI PTSFQARQLI LQYGLTLSDL DRHPEIDLAI DGADEVDADL NLIKGGGGCL TQEKIVAGYA SRFIVIADFR KDSKNLGDQW HKGIPIEVIP MAYVPVSRAV SQKFGGVVEL RMAVNKAGPV VTDNGNFILD WKFDRVHKWS EVNTAIKMIP GVVDTGLFIN MAERVYFGMQ DGSVNMREKP FC (1-311 aa encoded by BC015529)
Defects in RPIA are the cause of ribose 5-phosphate isomerase deficiency. The exact function of RPIA remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Phosphoriboisomerase; ribose 5-phosphate epimerase; Ribose-5-phosphate isomerase
Gene Aliases: RPI; RPIA; RPIAD
UniProt ID: (Human) P49247, (Mouse) P47968
Entrez Gene ID: (Human) 22934, (Mouse) 19895, (Rat) 362383
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support