Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RPL10 shares 98% amino acid sequence with RPL10L, so RPL10 antibody is able to recognize RPL10 and RPL10L protein.
Immunogen sequence: MGRRPARCY RYCKNKPYPK SRFCRGVPDA KIRIFDLGRK KAKVDEFPLC GHMVSDEYEQ LSSEALEAAR ICANKYMVKS CGKDGFHIRV RLHPFHVIRI NKMLSCAGAD RLQTGMRGAF GKPQGTVARV HIGQVIMSIR TKLQNKEHVI EALRRAKFKF PGRQKIHISK KWGFTKFNAD EFEDMVAEKR LIPDGCGVKY IPSRGPLDKW RALHS (1-214 aa encoded by BC003358)
This gene encodes a ribosomal protein that is a component of the 60S ribosome subunit. The related protein in chicken can bind to c-Jun and can repress c-Jun-mediated transcriptional activation. Some studies have detected an association between variation in this gene and autism spectrum disorders, though others do not detect this relationship. There are multiple pseudogenes of this gene dispersed throughout the genome. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 24.6 kDa protein; 24.6kDa protein; 60S ribosomal protein L10; Laminin receptor homolog; Large ribosomal subunit protein uL16; Protein QM; Protein QM homolog; ribosomal protein 10; Ribosomal protein L10; RPLPO; Tumor suppressor QM; Wilms tumor-related protein
Gene Aliases: AUTSX5; D0HXS648; DXHXS648; DXHXS648E; DXS648; DXS648E; L10; NOV; QM; QM16; RPL10
UniProt ID: (Human) P27635, (Rat) Q6PDV7, (Mouse) Q6ZWV3
Entrez Gene ID: (Human) 6134, (Rat) 81764, (Mouse) 110954
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support