Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVFRRFVEV GRVAYVSFGP HAGKLVAIVD VIDQNRALVD GPCTQVRRQA MPFKCMQLTD FILKFPHSAH QKYVRQAWQK ADINTKWAAT RWAKKIEARE RKAKMTDFDR FKVMKAKKMR NRIIKNEVKK LQKAALLKAS PKKAPGTKGT AAAAAAAAAA AKVPAKKITA ASKKAPAQKV PAQKATGQKA APAPKAQKGQ KAPAQKAPAP KASGKKA (1-216 aa encoded by BC000606)
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L14E family of ribosomal proteins. It contains a basic region-leucine zipper (bZIP)-like domain. The protein is located in the cytoplasm. This gene contains a trinucleotide (GCT) repeat tract whose length is highly polymorphic; these triplet repeats result in a stretch of alanine residues in the encoded protein. Transcript variants utilizing alternative polyA signals and alternative 5'-terminal exons exist but all encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Aoki M., Diabetes 45:157-164(1996). Tanaka M., Biochem. Biophys. Res. Commun. 243:531-537(1998). Yoshihama M., Genome Res. 12:379-390(2002).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L14; CAG-ISL 7; Large ribosomal subunit protein eL14; MGC88594
Gene Aliases: 3100001N19Rik; AA407502; AL022816; CAG-ISL-7; CTG-B33; hRL14; L14; RL14; RPL14
UniProt ID: (Human) P50914, (Mouse) Q9CR57
Entrez Gene ID: (Human) 9045, (Rat) 65043, (Mouse) 67115
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support