Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FLSKRNPRQI NWTVLYRRKH KKGQSEEIQK KRTRRAVKFQ RAITGASLAD I
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L24E family of ribosomal proteins. It is located in the cytoplasm. This gene has been referred to as ribosomal protein L30 because the encoded protein shares amino acid identity with the L30 ribosomal proteins from S. cerevisiae; however, its official name is ribosomal protein L24. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L24; 60S ribosomal protein L30; epididymis secretory protein Li 310; Large ribosomal subunit protein eL24; ribosomal protein L30
Gene Aliases: HEL-S-310; L24; RPL24
UniProt ID: (Human) P83731
Entrez Gene ID: (Human) 6152
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support