Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEGVEEKKK EVPAVPETLK KKRRNFAELK IKRLRKKFAQ KMLRKARRKL IYEKAKHYHK EYRQMYRTEI RMARMARKAG NFYVPAEPKL AFVIRIRGIN GVSPKVRKVL QLLRLRQIFN GTFVKLNKAS INMLRIVEPY IAWGYPNLKS VNELIYKRGY GKINKKRIAL TDNALIARSL GKYGIICMED LIHEIYTVGK RFKEANNFLW PFKLSSPRGG MKKKTTHFVE GGDAGNREDQ INRLIRRMN (1-248 aa encoded by BC006095 )
NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L7; Large ribosomal subunit protein uL30
Gene Aliases: humL7-1; L7; RPL7; Rpl7a; Surf-3
UniProt ID: (Human) P18124, (Mouse) P14148
Entrez Gene ID: (Human) 6129, (Mouse) 19989, (Rat) 297755
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support