Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GYGKINKKRI ALTDNALIAR SLGKYGIICM EDLIHEIYTV GKRFKEANNF LWPFKLSSPR GGMKKKTTHF VEGGDAGNRE DQINRLIRRM N
NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L7; Large ribosomal subunit protein uL30
Gene Aliases: humL7-1; L7; RPL7
UniProt ID: (Human) P18124
Entrez Gene ID: (Human) 6129
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support