Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: 293T, PC-12, HepG2, NIH/3T3, Raji, Mouse lung, Mouse liver, Rat lung, Rat liver
Immunogen sequence: MPKGKKAKGK KVAPAPAVVK KQEAKKVVNP LFEKRPKNFG IGQDIQPKRD LTRFVKWPRY IRLQRQRAIL YKRLKVPPAI NQFTQALDRQ TATQLLKLAH KYRPETKQEK KQRLLARAEK KAAGKGDVPT KRPPVLRAGV NTVTTLVENK KAQLVVIAHD VDPIE
Cytoplasmic ribosomes, organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L7AE family of ribosomal proteins. It can interact with a subclass of nuclear hormone receptors, including thyroid hormone receptor, and inhibit their ability to transactivate by preventing their binding to their DNA response elements. This gene is included in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. It is co-transcribed with the U24, U36a, U36b, and U36c small nucleolar RNA genes, which are located in its second, fifth, fourth, and sixth introns, respectively. This gene rearranges with the trk proto-oncogene to form the chimeric oncogene trk-2h, which encodes an oncoprotein consisting of the N terminus of ribosomal protein L7a fused to the receptor tyrosine kinase domain of trk. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L7a; Large ribosomal subunit protein eL8; PLA-X polypeptide; surfeit 3; Surfeit locus protein 3; thyroid hormone receptor uncoupling protein
Gene Aliases: L7A; RPL7A; SURF-3; SURF3; TRUP
UniProt ID: (Human) P62424, (Mouse) P12970, (Rat) P62425
Entrez Gene ID: (Human) 6130, (Mouse) 27176, (Rat) 296596
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support