Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAGPVKDRE AFQRLNFLYQ AAHCVLAQDP ENQALARFYC YTERTIAKRL VLRRDPSVKR TLCRGCSSLL VPGLTCTHRQ RRCRGQRWTV QTCLTCQRSQ RFLNDPGHLL WGDRPEAQLG SQADSKPLQP LPNTAHSISD RLPEEKMQTQ GSSNQ (1-154 aa encoded by BC011730)
RPP21 is a protein subunit of nuclear ribonuclease P, which processes the 5-prime leader sequence of precursor tRNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ribonuclease P 21 subunit; ribonuclease P 21kDa subunit; Ribonuclease P protein subunit p21; Ribonuclease P/MRP 21 kDa subunit; ribonuclease P/MRP 21kDa subunit; Ribonucleoprotein V; RNaseP protein p21
Gene Aliases: 0610037N01Rik; C6orf135; CAT60; Flj22638; RPP21
UniProt ID: (Human) Q9H633, (Mouse) Q8R040
Entrez Gene ID: (Human) 79897, (Mouse) 67676, (Rat) 406230
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support