Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MENFRKVRS EEAPAGCGAE GGGPGSGPFA DLAPGAVHMR VKEGSKIRNL MAFATASMAQ PATRAIVFSG CGRATTKTVT CAEILKRRLA GLHQVTRLRY RSVREVWQSL PPGPTQGQTP GEPAASLSVL KNVPGLAILL SKDALDPRQP GYQPPNPHPG PSSPPAAPAS KRSLGEPAAG EGSAKRSQPE PGVADEDQTA (1-199 aa encoded by BC002497)
Component of ribonuclease P, a ribonucleoprotein complex that generates mature tRNA molecules by cleaving their 5'-ends (PubMed:12003489, PubMed:16723659, PubMed:30454648). Also a component of the MRP ribonuclease complex, which cleaves pre-rRNA sequences (PubMed:28115465). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ribonuclease P 25 subunit; ribonuclease P 25kDa subunit; Ribonuclease P protein subunit p25; ribonuclease P/MRP 25kDa subunit; RNase P protein subunit p25
Gene Aliases: AI851155; RPP25
UniProt ID: (Human) Q9BUL9, (Rat) Q5PPN2, (Mouse) Q91WE3
Entrez Gene ID: (Human) 54913, (Rat) 315705, (Mouse) 102614
Molecular Function:
endoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support