Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VVKTSLNNPY IIRWSALESE DMHFILQTLE DRLKAIGLQK IEDKKKKNKT PFLKKESREK CSIAVDISEN LKEKKTDAKQ QVSGWTPAHV R
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. RPP38 may associate transiently with RNase P RNA as a factor involved in the transport of H1 RNA to the putative site of its assembly in the cell, the nucleolus.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ribonuclease P protein subunit p38; ribonuclease P/MRP 38kDa subunit; RNaseP protein p38
Gene Aliases: RPP38
UniProt ID: (Human) P78345
Entrez Gene ID: (Human) 10557
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support