Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAEVEQKKK RTFRKFTYRG VDLDQLLDMS YEQLMQLYSA RQRRRLNRGL RRKQHSLLKR LRKAKKEAPP MEKPEVVKTH LRDMIILPEM VGSMVGVYNG KTFNQVEIKP EMIGHYLGEF SITYKPVKHG RPGIGATHSS RFIPLK (1-145 aa encoded by B C064908)
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S19P family of ribosomal proteins. It is located in the cytoplasm. This gene has been found to be activated in various tumors, such as insulinomas, esophageal cancers, and colon cancers. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S15; homolog of rat insulinoma; insulinoma; insulinoma protein; RIG protein; Small ribosomal subunit protein uS19
Gene Aliases: RIG; RPS15; S15
UniProt ID: (Human) P62841, (Rat) P62845, (Mouse) P62843
Entrez Gene ID: (Human) 6209, (Rat) 29285, (Mouse) 20054
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support