Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPGVTVKDV NQQEFVRALA AFLKKSGKLK VPEWVDTVKL AKHKELAPYD ENWFYTRAAS TARHLYLRGG AGVGSMTKIY GGRQRNGVMP SHFSRGSKSV ARRVLQALEG LKMVEKDQDG GRKLTPQGQR DLDRIAGQVA AANKKH (1-145 aa encoded by B C000023)
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S19E family of ribosomal proteins. It is located in the cytoplasm. Mutations in this gene cause Diamond-Blackfan anemia, a constitutional erythroblastopenia characterized by absent or decreased erythroid precursors, in a subset of patients. This suggests a possible extra-ribosomal function for this gene in erythropoietic differentiation and proliferation, in addition to its ribosomal function. Higher expression levels of this gene in some primary colon carcinomas compared to matched normal colon tissues has been observed. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S19; HGNC:10402; Small ribosomal subunit protein eS19
Gene Aliases: DBA; DBA1; Dsk3; RPS19; S19
UniProt ID: (Human) P39019, (Rat) P17074, (Mouse) Q9CZX8
Entrez Gene ID: (Human) 6223, (Rat) 29287, (Mouse) 20085
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support